Assay Details
Gene Symbol: TEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754330.3 | XP_002754376.2 | 1 - 2 | 686 | 64 | |
XM_017977393.1 | XP_017832882.1 | 1 - 2 | 690 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
