Assay Details
Gene Symbol: TEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754330.3 | XP_002754376.2 | 2 - 3 | 762 | 76 | |
XM_017977393.1 | XP_017832882.1 | 2 - 3 | 766 | 76 | ||
XM_017977394.1 | XP_017832883.1 | 2 - 3 | 194 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017977394.1 transcripts.
Set Membership: |
![]() ![]() |
