Assay Details
Gene Symbol: TEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754330.3 | XP_002754376.2 | 3 - 4 | 961 | 111 | |
XM_017977393.1 | XP_017832882.1 | 3 - 4 | 965 | 111 | ||
XM_017977394.1 | XP_017832883.1 | 3 - 4 | 393 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
