Assay Details
Gene Symbol: TEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754330.3 | XP_002754376.2 | 4 - 5 | 1119 | 61 | |
XM_017977393.1 | XP_017832882.1 | 4 - 5 | 1123 | 61 | ||
XM_017977394.1 | XP_017832883.1 | 4 - 5 | 551 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
