Assay Details
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752160.3 | XP_002752206.1 | 1 - 2 | 285 | 83 | |
XM_002752162.3 | XP_002752208.1 | 1 - 2 | 285 | 83 | ||
XM_009003341.2 | XP_009001589.1 | 1 - 2 | 285 | 83 | ||
XM_009003342.2 | XP_009001590.1 | 1 - 2 | 285 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
