Assay Details
Gene Symbol: LOC100402350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749408.4 | XP_002749454.1 | 37 - 38 | 5451 | 111 | |
XM_008998829.2 | XP_008997077.1 | 4 - 5 | 574 | 111 | ||
XM_008998830.2 | XP_008997078.1 | 3 - 4 | 393 | 111 |

More Information
Set Membership: |
![]() ![]() |
