Assay Details
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754671.4 | XP_002754717.1 | 9 - 10 | 1403 | 73 | |
XM_003734146.3 | XP_003734194.1 | 9 - 10 | 1198 | 73 | ||
XM_009007222.2 | XP_009005470.1 | 9 - 10 | 1198 | 73 | ||
XM_009007224.2 | XP_009005472.1 | 9 - 10 | 1198 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
