Assay Details
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008994096.2 | XP_008992344.1 | 3 - 4 | 457 | 104 | |
XM_008994097.2 | XP_008992345.1 | 4 - 5 | 423 | 104 | ||
XM_008994098.2 | XP_008992346.1 | 4 - 5 | 375 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
