Assay Details
| Gene Symbol: LOC100385253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753603.2 | XP_002753649.2 | 4 - 5 | 1101 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100385253| Gene Symbol: LOC100385253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753603.2 | XP_002753649.2 | 4 - 5 | 1101 | 57 | |
Set Membership: |
|