Assay Details
Gene Symbol: NSUN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750774.3 | XP_002750820.1 | 3 - 4 | 1033 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
