Assay Details
Gene Symbol: ETAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002757685.3 | XP_002757731.1 | 5 - 6 | 2682 | 109 | |
XM_017964263.1 | XP_017819752.1 | 3 - 4 | 2223 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
