Assay Details
Gene Symbol: ZNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008982965.2 | XP_008981213.1 | 4 - 5 | 395 | 105 | |
XM_008982966.2 | XP_008981214.1 | 4 - 5 | 438 | 105 | ||
XM_017965476.1 | XP_017820965.1 | 4 - 5 | 463 | 105 | ||
XM_017965478.1 | XP_017820967.1 | 4 - 5 | 430 | 105 | ||
XM_017965480.1 | XP_017820969.1 | 3 - 4 | 398 | 105 | ||
XM_017965481.1 | XP_017820970.1 | 3 - 4 | 749 | 105 | ||
XM_017965484.1 | XP_017820973.1 | 3 - 4 | 235 | 105 | ||
XM_017965485.1 | XP_017820974.1 | 2 - 3 | 207 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017965484.1, XM_017965485.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
