Assay Details
Gene Symbol: POPDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758774.3 | XP_002758820.1 | 1 - 2 | 668 | 78 | |
XM_008982389.2 | XP_008980637.1 | 1 - 2 | 668 | 78 | ||
XM_008982390.2 | XP_008980638.1 | 1 - 2 | 668 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
