Assay Details
Gene Symbol: DKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260481.1 | NP_001247410.1 | 2 - 3 | 588 | 93 | |
GenBank mRNA | JN966749.1 | - | 2 - 3 | 588 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
