Assay Details
| Gene Symbol: DKK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260481.1 | NP_001247410.1 | 3 - 4 | 731 | 110 | |
| GenBank mRNA | JN966749.1 | - | 3 - 4 | 731 | 110 | |
More Information
Set Membership: |
|

DKK1
Genomic Map