Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991601.2 | XP_008989849.1 | 11 - 12 | 1237 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRAMD2B| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991601.2 | XP_008989849.1 | 11 - 12 | 1237 | 110 | |
Set Membership: |
|