Assay Details
Gene Symbol: PRRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760377.4 | XP_002760423.1 | 2 - 3 | 1566 | 69 | |
XM_008985042.2 | XP_008983290.1 | 2 - 3 | 1582 | 69 | ||
XM_008985043.2 | XP_008983291.1 | 2 - 3 | 1585 | 69 | ||
XM_008985044.2 | XP_008983292.1 | 2 - 3 | 1569 | 69 | ||
XM_017966095.1 | XP_017821584.1 | 4 - 5 | 693 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
