Assay Details
| Gene Symbol: BLVRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762136.3 | XP_002762182.1 | 1 - 2 | 234 | 61 | |
| XM_017967855.1 | XP_017823344.1 | 1 - 2 | 172 | 61 | ||
More Information
Set Membership: |
|

BLVRB
Genomic Map