Assay Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017976458.1 | XP_017831947.1 | 2 - 3 | 1479 | 73 | |
XM_017976460.1 | XP_017831949.1 | 2 - 3 | 1511 | 73 | ||
XM_017976461.1 | XP_017831950.1 | 2 - 3 | 1507 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
