Assay Details
Gene Symbol: LOC100398238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008998106.2 | XP_008996354.1 | 1 - 2 | 711 | 93 | |
XM_008998108.2 | XP_008996356.1 | 1 - 2 | 711 | 93 | ||
XM_008998109.2 | XP_008996357.1 | 1 - 2 | 711 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008998106.2, XM_008998108.2, XM_008998109.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
