Assay Details
| Gene Symbol: PDZRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965028.1 | XP_017820517.1 | 5 - 6 | 3707 | 60 | |
| XM_017965029.1 | XP_017820518.1 | 5 - 6 | 5257 | 60 | ||
| XM_017965030.1 | XP_017820519.1 | 5 - 6 | 12048 | 60 | ||
| XM_017965031.1 | XP_017820520.1 | 6 - 7 | 712 | 60 | ||
More Information
Set Membership: |
|

PDZRN3
Genomic Map