Assay Details
| Gene Symbol: RSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761452.2 | XP_002761498.1 | 9 | 1315 | 107 | |
| XM_017967224.1 | XP_017822713.1 | 9 | 1204 | 107 | ||
More Information
Set Membership: |
|

RSPH1
Genomic Map