Assay Details
| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988219.2 | XP_008986467.1 | 5 - 6 | 490 | 96 | |
| XM_008988220.2 | XP_008986468.1 | 4 - 5 | 407 | 96 | ||
| XM_008988221.2 | XP_008986469.1 | 4 - 5 | 426 | 96 | ||
More Information
Set Membership: |
|

TRAPPC6A
Genomic Map