Assay Details
Gene Symbol: LNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745839.4 | XP_002745885.1 | 6 - 7 | 1492 | 65 | |
XM_002745840.4 | XP_002745886.1 | 4 - 5 | 1047 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
