Assay Details
Gene Symbol: GORAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003735199.3 | XP_003735247.2 | 4 - 5 | 753 | 61 | |
XM_017966096.1 | XP_017821585.1 | 4 - 5 | 761 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
