Assay Details
Gene Symbol: FYB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745030.4 | XP_002745076.1 | 4 - 5 | 4126 | 122 | |
XM_003732356.3 | XP_003732404.1 | 4 - 5 | 2353 | 122 | ||
XM_008992136.2 | XP_008990384.1 | 4 - 5 | 4125 | 122 | ||
XM_008992137.2 | XP_008990385.1 | 4 - 5 | 4127 | 122 | ||
XM_008992139.2 | XP_008990387.1 | 4 - 5 | 4127 | 122 | ||
XM_008992140.2 | XP_008990388.1 | 4 - 5 | 4126 | 122 | ||
XM_017969940.1 | XP_017825429.1 | 4 - 5 | 2668 | 122 | ||
XM_017969942.1 | XP_017825431.1 | 4 - 5 | 1914 | 122 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
