Assay Details
Gene Symbol: CPLANE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750350.3 | XP_002750396.2 | 5 - 6 | 675 | 66 | |
XM_017974358.1 | XP_017829847.1 | 5 - 6 | 712 | 66 | ||
XM_017974359.1 | XP_017829848.1 | 4 - 5 | 573 | 66 | ||
XM_017974360.1 | XP_017829849.1 | 4 - 5 | 597 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
