Assay Details
Gene Symbol: HMGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745215.4 | XP_002745261.1 | 4 - 5 | 1453 | 104 | |
XM_008992446.2 | XP_008990694.1 | 4 - 5 | 1479 | 104 | ||
XM_008992447.2 | XP_008990695.1 | 5 - 6 | 806 | 104 | ||
XM_008992448.2 | XP_008990696.1 | 5 - 6 | 847 | 104 | ||
XM_008992449.2 | XP_008990697.1 | 4 - 5 | 780 | 104 | ||
XM_008992450.2 | XP_008990698.1 | 5 - 6 | 738 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
