Assay Details
| Gene Symbol: TUBB2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970942.1 | XP_017826431.1 | 1 - 2 | 118 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017970942.1 transcripts.
Set Membership: |
|

TUBB2B
Genomic Map