Assay Details
| Gene Symbol: LOC100395388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742885.4 | XP_002742931.1 | 2 - 3 | 271 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100395388| Gene Symbol: LOC100395388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742885.4 | XP_002742931.1 | 2 - 3 | 271 | 90 | |
Set Membership: |
|