Assay Details
| Gene Symbol: COQ5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753088.3 | XP_002753134.1 | 1 - 2 | 240 | 96 | |
| XM_009004776.2 | XP_009003024.1 | 1 - 2 | 240 | 96 | ||
| XM_009004777.2 | XP_009003025.1 | 1 - 2 | 240 | 96 | ||
| XM_009004779.2 | XP_009003027.1 | 1 - 2 | 240 | 96 | ||
More Information
Set Membership: |
|

COQ5
Genomic Map