Assay Details
Gene Symbol: WEE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002807320.3 | XP_002807366.1 | 4 - 5 | 922 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
