Assay Details
Gene Symbol: HCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017967672.1 | XP_017823161.1 | 1029 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
