Assay Details
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756717.3 | XP_002756763.1 | 3 - 4 | 390 | 81 | |
XM_008978913.2 | XP_008977161.1 | 4 - 5 | 489 | 81 | ||
XM_017963276.1 | XP_017818765.1 | 4 - 5 | 574 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
