Assay Details
Gene Symbol: FAM110A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747430.3 | XP_002747476.1 | 2 | 1327 | 106 | |
XM_008995611.1 | XP_008993859.1 | 2 | 772 | 106 | ||
XM_008995612.2 | XP_008993860.1 | 3 | 759 | 106 | ||
XM_008995615.2 | XP_008993863.1 | 2 | 565 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
