Assay Details
| Gene Symbol: MAPK1IP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753958.4 | XP_002754004.1 | 4 | 2674 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAPK1IP1L| Gene Symbol: MAPK1IP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753958.4 | XP_002754004.1 | 4 | 2674 | 105 | |
Set Membership: |
|