Assay Details
| Gene Symbol: TIGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008465.2 | XP_009006713.1 | 2 | 375 | 114 | |
| XM_009008466.2 | XP_009006714.1 | 2 | 436 | 114 | ||
More Information
Set Membership: |
|

TIGD3
Genomic Map