Assay Details
| Gene Symbol: SPIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989381.2 | XP_008987629.1 | 2 | 474 | 62 | |
| XM_008989382.2 | XP_008987630.1 | 2 | 447 | 62 | ||
More Information
Set Membership: |
|

SPIN3
Genomic Map