Assay Details
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747723.4 | XP_002747769.1 | 6 | 2296 | 104 | |
XM_017972140.1 | XP_017827629.1 | 6 | 2242 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
