Assay Details
Gene Symbol: ANAPC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199361.2 | NP_001186290.1 | 4 - 5 | 346 | 108 | |
NM_001282057.1 | NP_001268986.1 | 4 - 5 | 350 | 108 | ||
NM_001282058.1 | NP_001268987.1 | 4 - 5 | 323 | 108 | ||
NM_001282059.1 | NP_001268988.1 | 3 - 4 | 266 | 108 | ||
XM_009007484.2 | XP_009005732.1 | 4 - 5 | 396 | 108 | ||
XM_009007485.2 | XP_009005733.1 | 4 - 5 | 401 | 108 | ||
GenBank mRNA | EH382019.1 | - | 3 - 4 | 236 | 108 | |
HX385092.1 | - | 4 - 5 | 323 | 108 | ||
HX397241.1 | - | 4 - 5 | 351 | 108 | ||
HX407456.1 | - | 4 - 5 | 347 | 108 | ||
HX429160.1 | - | 4 - 5 | 315 | 108 | ||
HX658112.1 | - | 3 - 4 | 210 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
