Assay Details
| Gene Symbol: LHCGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301843.1 | NP_001288772.1 | 8 - 9 | 883 | 63 | |
| GenBank mRNA | U80673.1 | - | 8 - 9 | 883 | 63 | |
More Information
Set Membership: |
|

LHCGR
Genomic Map