Assay Details
| Gene Symbol: ARHGAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204391.1 | NP_001191320.1 | 8 - 9 | 749 | 58 | |
| XM_017971223.1 | XP_017826712.1 | 8 - 9 | 816 | 58 | ||
| XM_017971438.1 | XP_017826927.1 | 7 - 8 | 716 | 58 | ||
| XM_017971633.1 | XP_017827122.1 | 8 - 9 | 816 | 58 | ||
| XM_017971871.1 | XP_017827360.1 | 8 - 9 | 816 | 58 | ||
| XM_017972125.1 | XP_017827614.1 | 8 - 9 | 816 | 58 | ||
More Information
Set Membership: |
|

ARHGAP8
Genomic Map