Assay Details
| Gene Symbol: SELENOW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278377.1 | NP_001265306.1 | 5 - 6 | 414 | 97 | |
| GenBank mRNA | HX387247.1 | - | 5 - 6 | 402 | 98 | |
More Information
Set Membership: |
|

SELENOW
Genomic Map