Assay Details
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281866.1 | NP_001268795.1 | 1 - 2 | 47 | 87 | |
XM_008996476.2 | XP_008994724.1 | 1 - 2 | 1329 | 87 | ||
GenBank mRNA | HX638469.1 | - | 1 - 2 | 48 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001281866.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
