Assay Details
| Gene Symbol: OAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290218.1 | NP_001277147.1 | 4 - 5 | 635 | 69 | |
| GenBank mRNA | HX649635.1 | - | 4 - 5 | 587 | 69 | |
More Information
Set Membership: |
|

OAZ3
Genomic Map