Assay Details
| Gene Symbol: NKX3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756836.2 | XP_002756882.1 | 1 - 2 | 297 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKX3-1| Gene Symbol: NKX3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756836.2 | XP_002756882.1 | 1 - 2 | 297 | 74 | |
Set Membership: |
|