Assay Details
| Gene Symbol: LOC100401187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002555.2 | XP_009000803.1 | 1 | 422 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100401187| Gene Symbol: LOC100401187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002555.2 | XP_009000803.1 | 1 | 422 | 84 | |
Set Membership: |
|