Assay Details
Gene Symbol: C8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750872.2 | XP_002750918.1 | 3 - 4 | 446 | 57 | |
XM_017974789.1 | XP_017830278.1 | 3 - 4 | 446 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
