Assay Details
Gene Symbol: ASTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002757375.2 | XP_002757421.1 | 1 - 2 | 57 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
