Assay Details
Gene Symbol: BDKRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754262.3 | XP_002754308.1 | 2 - 3 | 263 | 108 | |
XM_009006516.2 | XP_009004764.1 | 2 - 3 | 274 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
